Assay Details
Target Gene Details
Entrez Gene ID: | 1142 |
Gene Name: | cholinergic receptor nicotinic beta 3 subunit |
Gene Aliases: |
- |
Location: |
Chr.8:42697419-42737407 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNB3 | NM_000749.3 | NP_000740.1 | ||
| BC069681.1 | AAH69681.1 | |||
| BC069703.1 | AAH69703.1 | |||
| BC069788.1 | AAH69788.1 | |||
| U62438.1 | AAB40116.1 | |||
| X67513.1 | CAA47851.1 | |||
| Y08417.1 | CAA69694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7186n100 | Chr.8:42688183 - 42763570 on Build GRCh38 | Gain |
|
| nsv1161758 | Chr.8:42666258 - 42733252 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map