Assay Details
Target Gene Details
Entrez Gene ID: | 55423 |
Gene Name: | signal regulatory protein gamma |
Gene Aliases: |
CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1 |
Location: |
Chr.20:1629152-1686516 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPG | NM_001039508.1 | 5 | 1005 | NP_001034597.1 |
| NM_018556.3 | 6 | 1338 | NP_061026.2 | |
| NM_080816.2 | 4 | 687 | NP_543006.2 | |
| XM_005260749.3 | 5 | 1056 | XP_005260806.1 | |
| XM_011529286.2 | 6 | 1385 | XP_011527588.1 | |
| AA446468.1 | ||||
| AB042624.1 | 6 | 1354 | BAA95692.1 | |
| AK124224.1 | 1 | 3718 | ||
| AK292745.1 | 6 | 1338 | ||
| BC020629.1 | 4 | 664 | ||
| BC064532.1 | 6 | 1299 | AAH64532.1 | |
| DB046293.1 | 3 | 371 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| nsv510779 | Chr.20:1528684 - 1633316 on Build GRCh38 | Deletion |
|
| esv2722074 | Chr.20:1604818 - 1636346 on Build GRCh38 | Deletion |
|
| esv2758777 | Chr.20:1496135 - 1698610 on Build GRCh38 | Gain+Loss |
|
| esv3644988 | Chr.20:1626418 - 1861080 on Build GRCh38 | Gain |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| esv2722071 | Chr.20:1565136 - 1694217 on Build GRCh38 | Deletion |
|
| nsv833894 | Chr.20:1538787 - 1772720 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map