Assay Details
Target Gene Details
Entrez Gene ID: | 114826 |
Gene Name: | SET and MYND domain containing 4 |
Gene Aliases: |
ZMYND21 |
Location: |
Chr.17:1779535-1829896 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMYD4 | NM_052928.2 | 11 | 4238 | NP_443160.2 |
| XM_017024121.1 | 12 | 4483 | XP_016879610.1 | |
| AL137345.1 | 1 | 1286 | ||
| BC035077.2 | 11 | 4157 | AAH35077.1 | |
| BM676597.1 | 1 | 112 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9490 | Chr.17:1653428 - 1828993 on Build GRCh38 | Loss |
|
| nsv1065585 | Chr.17:1733183 - 1918769 on Build GRCh38 | Gain |
|
| nsv1946 | Chr.17:1717490 - 1781570 on Build GRCh38 | Insertion |
|
| nsv833335 | Chr.17:1713955 - 1827844 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map