Assay Details
Target Gene Details
Entrez Gene ID: | 5582 |
Gene Name: | protein kinase C gamma |
Gene Aliases: |
PKC-gamma, PKCC, PKCG, SCA14 |
Location: |
Chr.19:53882184-53907647 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCG | NM_001316329.1 | 6 | 948 | NP_001303258.1 |
| NM_002739.4 | 6 | 948 | NP_002730.1 | |
| AB209671.1 | 6 | 667 | BAD92908.1 | |
| AF345987.1 | 6 | 832 | AAK13533.1 | |
| AK296412.1 | 6 | 956 | ||
| AK296434.1 | 6 | 935 | ||
| AK302958.1 | 6 | 933 | ||
| AK303741.1 | 4 | 355 | ||
| AK312302.1 | 6 | 910 | ||
| BC047876.1 | 6 | 823 | AAH47876.1 | |
| M13977.1 | 6 | 832 | AAA60102.1 | |
| Z15114.1 | 2 | 156 | CAA78820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064558 | Chr.19:53773468 - 53957491 on Build GRCh38 | Gain |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
| nsv833876 | Chr.19:53845956 - 53995117 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map