Assay Details
Target Gene Details
Entrez Gene ID: | 283189 |
Gene Name: | olfactory receptor family 9 subfamily G member 4 |
Gene Aliases: |
OR11-216 |
Location: |
Chr.11:56742828-56743811 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR9G4 | NM_001005284.1 | 1 | 175 | NP_001005284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555101 | Chr.11:56736252 - 56936567 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map