Assay Details
Target Gene Details
Entrez Gene ID: | 100129128 |
Gene Name: | KH domain containing 1 like |
Gene Aliases: |
- |
Location: |
Chr.6:73223544-73225452 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHDC1L | NM_001126063.2 | NP_001119535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732286 | Chr.6:73210758 - 73243312 on Build GRCh38 | Deletion |
|
| nsv1030079 | Chr.6:73210327 - 73274361 on Build GRCh38 | Gain |
|
| esv3609501 | Chr.6:73223489 - 73233695 on Build GRCh38 | Gain |
|
| esv2732287 | Chr.6:73218973 - 73234353 on Build GRCh38 | Deletion |
|

Genomic Map