Assay Details
Target Gene Details
Entrez Gene ID: | 3820 |
Gene Name: | killer cell lectin like receptor B1 |
Gene Aliases: |
CD161, CLEC5B, NKR, NKR-P1, NKR-P1A, NKRP1A, hNKR-P1A |
Location: |
Chr.12:9595274-9607901 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRB1 | NM_002258.2 | 4 | 360 | NP_002249.1 |
| AK292022.1 | 4 | 375 | ||
| BC113997.1 | 4 | 348 | AAI13998.1 | |
| BC114516.1 | 4 | 348 | AAI14517.1 | |
| BF432813.1 | 4 | 361 | ||
| U11276.1 | 4 | 358 | AAA21605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| esv2759878 | Chr.12:9218999 - 9770933 on Build GRCh38 | Gain+Loss |
|
| nsv428271 | Chr.12:9407491 - 9678072 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map