Assay Details
Target Gene Details
Entrez Gene ID: | 93663 |
Gene Name: | Rho GTPase activating protein 18 |
Gene Aliases: |
MacGAP, SENEX, bA307O14.2 |
Location: |
Chr.6:129576145-129710225 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP18 | NM_033515.2 | 15 | 2886 | NP_277050.2 |
| XM_005267212.2 | 15 | 2962 | XP_005267269.2 | |
| XM_005267213.1 | 14 | 2761 | XP_005267270.1 | |
| XM_017011494.1 | 14 | 2837 | XP_016866983.1 | |
| AB053293.1 | 14 | 2684 | BAB61887.1 | |
| AK002195.1 | 2 | 924 | ||
| AL834511.1 | 15 | 2824 | CAD39167.2 | |
| AY643712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
|
| nsv524782 | Chr.6:129575356 - 129724532 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map