Assay Details
Target Gene Details
Entrez Gene ID: | 9839 |
Gene Name: | zinc finger E-box binding homeobox 2 |
Gene Aliases: |
HSPC082, SIP-1, SIP1, SMADIP1, ZFHX1B |
Location: |
Chr.2:144384375-144520391 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZEB2 | NM_001171653.1 | 7 | 2776 | NP_001165124.1 |
| NM_014795.3 | 8 | 2848 | NP_055610.1 | |
| XM_006712881.3 | 8 | 2442 | XP_006712944.1 | |
| XM_006712882.3 | 8 | 2491 | XP_006712945.1 | |
| XM_011512232.2 | 7 | 2485 | XP_011510534.1 | |
| XM_017005414.1 | 9 | 2562 | XP_016860903.1 | |
| XM_017005415.1 | 7 | 3465 | XP_016860904.1 | |
| AB011141.1 | 8 | 2770 | ||
| AB056507.1 | 8 | 2805 | BAB40819.1 | |
| AB193096.1 | 7 | 2326 | BAD60928.1 | |
| AK294928.1 | 7 | 2504 | ||
| BC127101.1 | 8 | 2460 | ||
| BC127102.1 | 8 | 2459 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592768 | Chr.2:144379711 - 144546411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map