Assay Details
Target Gene Details
Entrez Gene ID: | 5588 |
Gene Name: | protein kinase C theta |
Gene Aliases: |
PRKCT, nPKC-theta |
Location: |
Chr.10:6393038-6580646 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCQ | NM_001242413.2 | 17 | 2512 | NP_001229342.1 |
| NM_001282644.1 | 18 | 2699 | NP_001269573.1 | |
| NM_001282645.1 | 17 | 2640 | NP_001269574.1 | |
| NM_001323265.1 | 18 | 2756 | NP_001310194.1 | |
| NM_001323266.1 | 17 | 2638 | NP_001310195.1 | |
| NM_001323267.1 | NP_001310196.1 | |||
| NM_006257.4 | 18 | 2701 | NP_006248.1 | |
| XM_005252496.4 | 18 | 2801 | XP_005252553.1 | |
| XM_005252497.4 | XP_005252554.1 | |||
| XM_006717465.3 | 20 | 5196 | XP_006717528.1 | |
| XM_017016410.1 | 19 | 5107 | XP_016871899.1 | |
| XM_017016411.1 | 19 | 5135 | XP_016871900.1 | |
| AI570478.1 | 1 | 598 | ||
| BX647657.1 | 17 | 2786 | ||
| L01087.1 | 18 | 2695 | AAA75571.1 | |
| L07032.1 | 17 | 2622 | AAA60101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053177 | Chr.10:6411345 - 6431152 on Build GRCh38 | Loss |
|
| nsv1161925 | Chr.10:6417656 - 6429854 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map