Assay Details
Target Gene Details
Entrez Gene ID: | 8099 |
Gene Name: | cyclin dependent kinase 2 associated protein 1 |
Gene Aliases: |
DOC1, DORC1, ST19, doc-1, p12DOC-1 |
Location: |
Chr.12:123260970-123272316 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK2AP1 | NM_004642.3 | NP_004633.1 | ||
| NR_073007.1 | ||||
| AF006484.1 | AAC77831.1 | |||
| DR759851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv884n106 | Chr.12:123271053 - 123272153 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map