Assay Details
Target Gene Details
Entrez Gene ID: | 2350 |
Gene Name: | folate receptor beta |
Gene Aliases: |
BETA-HFR, FBP, FBP/PL-1, FR-BETA, FR-P3 |
Location: |
Chr.11:72216775-72221950 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOLR2 | NM_000803.4 | 5 | 1008 | NP_000794.3 |
| NM_001113534.1 | 5 | 1002 | NP_001107006.1 | |
| NM_001113535.1 | 5 | 996 | NP_001107007.1 | |
| NM_001113536.1 | 5 | 990 | NP_001107008.1 | |
| AK222539.1 | 5 | 999 | BAD96259.1 | |
| AK222643.1 | 5 | 990 | BAD96363.1 | |
| BC058036.1 | 5 | 992 | AAH58036.1 | |
| BC115366.1 | ||||
| CD014063.1 | 4 | 859 |
More Information
Set Membership: |
|

Genomic Map