Assay Details
Target Gene Details
Entrez Gene ID: | 84101 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
- |
Location: |
Chr.12:95516560-95551529 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP44 | NM_001042403.2 | 6 | 3275 | NP_001035862.1 |
| NM_001278393.1 | 6 | 3491 | NP_001265322.1 | |
| NM_032147.4 | 6 | 3420 | NP_115523.2 | |
| NR_125360.1 | 4 | 3005 | ||
| XM_005269172.1 | 7 | 3507 | XP_005269229.1 | |
| XM_005269173.1 | 7 | 3584 | XP_005269230.1 | |
| XM_005269174.1 | 7 | 3351 | XP_005269231.1 | |
| XM_011538800.2 | 8 | 3534 | XP_011537102.1 | |
| XM_011538801.1 | 8 | 3394 | XP_011537103.1 | |
| XM_011538802.1 | 7 | 3354 | XP_011537104.1 | |
| XM_011538803.1 | 7 | 3359 | XP_011537105.1 | |
| XM_011538804.2 | 7 | 3567 | XP_011537106.1 | |
| XM_011538805.1 | 7 | 4208 | XP_011537107.1 | |
| XM_017020009.1 | 6 | 3409 | XP_016875498.1 | |
| XM_017020010.1 | 6 | 3261 | XP_016875499.1 | |
| XM_017020011.1 | 7 | 3287 | XP_016875500.1 |
More Information
Set Membership: |
|

Genomic Map