Assay Details
Target Gene Details
Entrez Gene ID: | 10640 |
Gene Name: | exocyst complex component 5 |
Gene Aliases: |
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P |
Location: |
Chr.14:57202476-57268899 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOC5 | NM_006544.3 | 18 | 3432 | NP_006535.1 |
| XM_005267272.3 | 18 | 3517 | XP_005267329.1 | |
| AL833745.1 | 8 | 2899 | CAH56255.1 | |
| BC041126.1 | 18 | 3398 | AAH41126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046422 | Chr.14:57198718 - 58129812 on Build GRCh38 | Gain |
|
| nsv977472 | Chr.14:57207222 - 57208909 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map