Assay Details
Target Gene Details
Entrez Gene ID: | 55585 |
Gene Name: | ubiquitin conjugating enzyme E2 Q1 |
Gene Aliases: |
GTAP, NICE-5, PRO3094, UBE2Q |
Location: |
Chr.1:154548575-154558644 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2Q1 | NM_017582.6 | NP_060052.3 | ||
| AK056388.1 | BAB71173.1 | |||
| AK293986.1 | ||||
| AY112698.1 | AAM60814.1 | |||
| AY948200.1 | AAY23342.1 | |||
| BC000848.1 | AAH00848.1 | |||
| BC061583.1 | AAH61583.1 | |||
| BC070158.1 | AAH70158.1 | |||
| BC087836.1 | AAH87836.1 | |||
| BC107057.2 | AAI07058.1 | |||
| BC148185.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map