Assay Details
Target Gene Details
Entrez Gene ID: | 79698 |
Gene Name: | zinc finger matrin-type 4 |
Gene Aliases: |
- |
Location: |
Chr.8:40530590-40897828 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMAT4 | NM_001135731.1 | 6 | 686 | NP_001129203.1 |
| NM_024645.2 | 7 | 914 | NP_078921.1 | |
| AK023904.1 | 7 | 914 | BAB14719.1 | |
| BC019598.1 | 6 | 672 | AAH19598.1 |
More Information
Set Membership: |
|

Genomic Map