Assay Details
Target Gene Details
Entrez Gene ID: | 3832 |
Gene Name: | kinesin family member 11 |
Gene Aliases: |
EG5, HKSP, KNSL1, MCLMR, TRIP5 |
Location: |
Chr.10:92593068-92655395 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF11 | NM_004523.3 | 2 | 470 | NP_004514.2 |
| AA857025.1 | 2 | 302 | ||
| AK314258.1 | 2 | 373 | ||
| BC126211.1 | 2 | 269 | ||
| BC136474.1 | 2 | 269 | ||
| DB078134.1 | 2 | 470 | ||
| U37426.1 | 2 | 252 | AAA86132.1 | |
| X85137.1 | 2 | 172 | CAA59449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046258 | Chr.10:92107212 - 92706349 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map