Assay Details
Target Gene Details
Entrez Gene ID: | 129450 |
Gene Name: | tRNA-yW synthesizing protein 5 |
Gene Aliases: |
C2orf60, hTYW5 |
Location: |
Chr.2:199928911-199955736 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TYW5 | NM_001039693.2 | NP_001034782.1 | ||
| NR_004862.1 | ||||
| NR_109905.1 | ||||
| NR_109906.1 | ||||
| NR_109907.1 | ||||
| XM_017003310.1 | XP_016858799.1 | |||
| AK095272.1 | BAC04517.1 | |||
| AK293064.1 | ||||
| AK298977.1 | ||||
| BC132835.1 | ||||
| BC136837.1 | ||||
| BX482572.1 | ||||
| BX647677.1 | ||||
| BX648265.1 | 2 | 4761 | ||
| CN261486.1 | ||||
| HY182974.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834510 | Chr.2:199852719 - 200052794 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map