Assay Details
Target Gene Details
Entrez Gene ID: | 79958 |
Gene Name: | DENN domain containing 1C |
Gene Aliases: |
FAM31C |
Location: |
Chr.19:6467204-6481808 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND1C | NM_001290331.1 | 21 | 1851 | NP_001277260.1 |
| NM_024898.3 | 23 | 1960 | NP_079174.2 | |
| XM_006722905.2 | 21 | 1704 | XP_006722968.2 | |
| XM_006722906.3 | 23 | 1889 | XP_006722969.1 | |
| XM_011528318.2 | 22 | 1880 | XP_011526620.1 | |
| AK026410.1 | 23 | 1939 | BAB15475.1 | |
| AK055820.1 | 12 | 1567 | ||
| AK225623.1 | 23 | 1939 | ||
| AK303226.1 | 21 | 1830 | ||
| AL713770.1 | 21 | 2080 | ||
| AL834293.1 | 6 | 561 | CAD38966.1 | |
| BC033437.1 | 23 | 1927 | AAH33437.1 | |
| BX647304.1 | 11 | 1590 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116930831] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map