Assay Details
Target Gene Details
Entrez Gene ID: | 3376 |
Gene Name: | isoleucyl-tRNA synthetase |
Gene Aliases: |
IARS1, ILERS, ILRS, IRS, PRO0785 |
Location: |
Chr.9:92210207-92293756 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IARS | NM_002161.5 | 33 | 3778 | NP_002152.2 |
| NM_013417.3 | 33 | 3948 | NP_038203.2 | |
| NR_073446.1 | 32 | 3621 | ||
| AB209234.1 | 33 | 3687 | BAD92471.1 | |
| AK293014.1 | 33 | 3705 | ||
| AK301272.1 | 21 | 2258 | ||
| AK308202.1 | ||||
| BC008318.1 | 11 | 1283 | AAH08318.2 | |
| BC065552.1 | 32 | 3464 | AAH65552.1 | |
| BX537417.1 | 33 | 3716 | CAD97659.1 | |
| BX537429.1 | 33 | 3718 | CAD97671.1 | |
| BX538109.1 | 32 | 3561 | CAD98022.1 | |
| BX647071.1 | 33 | 3717 | ||
| BX647236.1 | 33 | 3710 | ||
| BX647430.1 | 33 | 3712 | ||
| BX648567.1 | 33 | 3718 | ||
| D28473.1 | 33 | 3866 | BAA05835.1 | |
| U04953.1 | 33 | 3880 | AAA80153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891693 | Chr.9:92163398 - 92538277 on Build GRCh38 | Gain |
|
| nsv1046733 | Chr.9:91928340 - 92376282 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map