Assay Details
Target Gene Details
Entrez Gene ID: | 8038 |
Gene Name: | ADAM metallopeptidase domain 12 |
Gene Aliases: |
ADAM12-OT1, CAR10, MCMP, MCMPMltna, MLTN, MLTNA |
Location: |
Chr.10:126012385-126388558 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM12 | NM_001288973.1 | 23 | 5169 | NP_001275902.1 |
| NM_003474.5 | 23 | 5178 | NP_003465.3 | |
| XM_017016705.1 | 22 | 5081 | XP_016872194.1 | |
| XM_017016706.1 | 13 | 4220 | XP_016872195.1 | |
| AF023476.2 | AAC08702.2 | |||
| KF444157.1 | 23 | 5169 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759788 | Chr.10:125765331 - 126098133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map