Assay Details
Target Gene Details
Entrez Gene ID: | 387 |
Gene Name: | ras homolog family member A |
Gene Aliases: |
ARH12, ARHA, RHO12, RHOH12 |
Location: |
Chr.3:49359136-49412097 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOA | NM_001313941.1 | 6 | 945 | NP_001300870.1 |
| NM_001313943.1 | 6 | 989 | NP_001300872.1 | |
| NM_001313944.1 | 5 | 790 | NP_001300873.1 | |
| NM_001313945.1 | 5 | 727 | NP_001300874.1 | |
| NM_001313946.1 | 3 | 598 | NP_001300875.1 | |
| NM_001313947.1 | 4 | 729 | NP_001300876.1 | |
| NM_001664.3 | 5 | 850 | NP_001655.1 | |
| AF498970.1 | AAM21117.1 | |||
| AK130066.1 | 5 | 846 | ||
| AK130808.1 | 5 | 726 | ||
| AK222556.1 | 5 | 742 | BAD96276.1 | |
| AK296654.1 | 5 | 666 | ||
| AK307220.1 | 5 | 625 | ||
| AW403464.1 | ||||
| BC000946.3 | 2 | 294 | AAH00946.2 | |
| BC001360.2 | 5 | 682 | AAH01360.1 | |
| BC005976.1 | 5 | 687 | AAH05976.1 | |
| BQ721697.1 | 6 | 862 | ||
| BT019870.1 | AAV38673.1 | |||
| BU509012.1 | 4 | 560 | ||
| BX647063.1 | 5 | 746 | CAE46190.1 | |
| L09159.1 | 4 | 559 | AAA50612.1 | |
| L25080.1 | 5 | 721 | AAC33178.1 | |
| LN607834.1 | 2 | 296 | ||
| X05026.1 | 5 | 728 | CAA28690.1 |
More Information
Set Membership: |
|

Genomic Map