Assay Details
Target Gene Details
Entrez Gene ID: | 7093 |
Gene Name: | tolloid like 2 |
Gene Aliases: |
- |
Location: |
Chr.10:96364606-96513926 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLL2 | NM_012465.3 | 21 | 4625 | NP_036597.1 |
| AB023149.2 | 21 | 4575 | ||
| AF059516.1 | 21 | 4748 | AAD42979.1 | |
| BC112341.1 | 21 | 4508 | AAI12342.1 | |
| BC112366.1 | 21 | 4508 | AAI12367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831950 | Chr.10:96321215 - 96485677 on Build GRCh38 | Gain |
|
| nsv1047657 | Chr.10:96363477 - 96423730 on Build GRCh38 | Loss |
|
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map