Assay Details
Target Gene Details
Entrez Gene ID: | 79092 |
Gene Name: | caspase recruitment domain family member 14 |
Gene Aliases: |
BIMP2, CARMA2, PRP, PSORS2, PSS1 |
Location: |
Chr.17:80169992-80209331 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD14 | NM_001257970.1 | 15 | 2329 | NP_001244899.1 |
| NM_024110.4 | 15 | 2329 | NP_077015.2 | |
| NR_047566.1 | 16 | 2326 | ||
| XM_011525212.1 | 17 | 2850 | XP_011523514.1 | |
| XM_011525213.1 | 17 | 2592 | XP_011523515.1 | |
| XM_011525215.1 | 16 | 2652 | XP_011523517.1 | |
| XM_011525216.1 | 16 | 2352 | XP_011523518.1 | |
| XM_011525217.1 | 16 | 2408 | XP_011523519.1 | |
| XM_011525218.2 | 16 | 2391 | XP_011523520.1 | |
| XM_017025057.1 | 17 | 2847 | XP_016880546.1 | |
| AF322642.1 | 15 | 2367 | AAG53403.1 | |
| AK002138.1 | ||||
| AK091123.1 | 16 | 2284 | ||
| AK296762.1 | 11 | 1423 | ||
| AX746790.1 | 16 | 2284 | ||
| AY032927.1 | 14 | 2164 | AAK54453.1 | |
| BC018142.1 | 15 | 2338 | AAH18142.1 | |
| EU652409.1 | 14 | 2161 |
Target Gene Details
Entrez Gene ID: | 6448 |
Gene Name: | N-sulfoglucosamine sulfohydrolase |
Gene Aliases: |
HSS, MPS3A, SFMD |
Location: |
Chr.17:80200668-80220400 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGSH |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066606 | Chr.17:79955666 - 80224006 on Build GRCh38 | Gain |
|
| nsv469591 | Chr.17:80085322 - 80293486 on Build GRCh38 | Loss |
|
| nsv576130 | Chr.17:80184196 - 80220032 on Build GRCh38 | Loss |
|
| nsv833562 | Chr.17:80114523 - 80301339 on Build GRCh38 | Loss |
|
| esv3641368 | Chr.17:80194137 - 80202540 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map