Assay Details
Target Gene Details
Entrez Gene ID: | 84075 |
Gene Name: | fibrous sheath CABYR binding protein |
Gene Aliases: |
C14orf155 |
Location: |
Chr.14:44504149-44507296 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSCB | NM_032135.3 | 1 | 1136 | NP_115511.3 |
| AK124110.1 | 1 | 1126 | ||
| AK302615.1 | 1 | 1090 | ||
| AL136775.1 | 1 | 463 | CAB66709.1 | |
| BC039878.1 | 1 | 1095 | AAH39878.1 | |
| CR933608.1 | 1 | 1119 | CAI45927.1 | |
| CR933626.1 | 1 | 1093 | CAI45935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564681 | Chr.14:44264601 - 44680343 on Build GRCh38 | Loss |
|
| esv3892524 | Chr.14:44070887 - 44781215 on Build GRCh38 | Loss |
|
| nsv456254 | Chr.14:43735368 - 44523056 on Build GRCh38 | Gain |
|
| nsv564642 | Chr.14:43382442 - 44942877 on Build GRCh38 | Loss |
|
| nsv456273 | Chr.14:44481683 - 44537275 on Build GRCh38 | Loss |
|
| esv3634329 | Chr.14:44426867 - 44536131 on Build GRCh38 | Loss |
|
| nsv1051850 | Chr.14:43222549 - 44531691 on Build GRCh38 | Loss |
|

Genomic Map