Assay Details
Target Gene Details
Entrez Gene ID: | 5266 |
Gene Name: | peptidase inhibitor 3 |
Gene Aliases: |
ESI, SKALP, WAP3, WFDC14, cementoin |
Location: |
Chr.20:45174899-45176544 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI3 | NM_002638.3 | NP_002629.1 | ||
| BC010952.1 | AAH10952.1 | |||
| CR542234.1 | CAG47030.1 | |||
| Z18538.1 | CAA79223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map