Assay Details
Target Gene Details
Entrez Gene ID: | 956 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 3 |
Gene Aliases: |
CD39L3, HB6, NTPDase-3 |
Location: |
Chr.3:40387156-40428626 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD3 | NM_001248.3 | NP_001239.2 | ||
| NM_001291960.1 | NP_001278889.1 | |||
| NM_001291961.1 | NP_001278890.1 | |||
| XM_011534266.2 | XP_011532568.1 | |||
| AF034840.2 | AAC09236.2 | |||
| AF039917.1 | AAC39884.1 | |||
| AK313322.1 | ||||
| BC029869.1 | AAH29869.1 |
Target Gene Details
Entrez Gene ID: | 285266 |
Gene Name: | ENTPD3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:40390951-40453308 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD3-AS1 | NR_040100.1 | |||
| BG527749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595950 | Chr.3:40411223 - 40420706 on Build GRCh38 | Loss |
|
| nsv590110 | Chr.3:40327497 - 40425488 on Build GRCh38 | Loss |
|
| esv2671271 | Chr.3:40411262 - 40420656 on Build GRCh38 | Deletion |
|
| esv3595947 | Chr.3:40342143 - 40444686 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map