Assay Details
Target Gene Details
Entrez Gene ID: | 265 |
Gene Name: | amelogenin, X-linked |
Gene Aliases: |
AI1E, AIH1, ALGN, AMG, AMGL, AMGX |
Location: |
Chr.X:11293413-11300761 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMELX | NM_001142.2 | 5 | 361 | NP_001133.1 |
| NM_182680.1 | 6 | 403 | NP_872621.1 | |
| NM_182681.1 | 4 | 313 | NP_872622.1 | |
| XM_017029404.1 | 4 | 1462 | XP_016884893.1 | |
| AF436849.1 | 6 | 403 | AAL30432.1 | |
| BC069118.1 | 5 | 361 | AAH69118.1 | |
| BC074951.2 | 5 | 340 | AAH74951.1 | |
| M86932.1 | 5 | 361 | AAA51717.1 | |
| S67147.1 | 1 | 149 | AAB29184.1 |
Target Gene Details
Entrez Gene ID: | 395 |
Gene Name: | Rho GTPase activating protein 6 |
Gene Aliases: |
RHOGAP6, RHOGAPX-1 |
Location: |
Chr.X:11137543-11665701 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP6 | NM_001287242.1 | NP_001274171.1 | ||
| NM_006125.2 | NP_006116.2 | |||
| NM_013427.2 | NP_038286.2 | |||
| NR_109776.1 | ||||
| AB208792.1 | BAD92029.1 | |||
| AF012272.2 | AAC98539.2 | |||
| AF022212.2 | AAC98540.2 | |||
| AF117067.1 | AAF43261.1 | |||
| AK293078.1 | ||||
| AK303444.1 | ||||
| BC150635.1 | ||||
| HY018118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739963 | Chr.X:11216833 - 11639506 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map