Assay Details
Target Gene Details
Entrez Gene ID: | 29927 |
Gene Name: | Sec61 translocon alpha 1 subunit |
Gene Aliases: |
HSEC61, SEC61, SEC61A |
Location: |
Chr.3:128052369-128071683 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC61A1 | NM_013336.3 | NP_037468.1 | ||
| AF077032.1 | AAD27765.1 | |||
| AF084458.1 | AAD39847.1 | |||
| AF346602.1 | AAK29083.1 | |||
| AK027423.1 | ||||
| AK074706.1 | ||||
| AK299118.1 | ||||
| AL832821.1 | CAI46127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001879 | Chr.3:127956099 - 128143493 on Build GRCh38 | Gain |
|
| nsv3991 | Chr.3:128048752 - 128081671 on Build GRCh38 | Insertion |
|
| nsv1002685 | Chr.3:127819453 - 128455596 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map