Assay Details
Target Gene Details
Entrez Gene ID: | 64215 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C1 |
Gene Aliases: |
DNAJL1, ERdj1, HTJ1, MTJ1 |
Location: |
Chr.10:21756548-22003768 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC1 | NM_022365.3 | NP_071760.2 | ||
| AF427142.1 | AAL24046.1 | |||
| AK026062.1 | BAB15343.1 | |||
| AK027263.1 | BAB55004.1 | |||
| AY225122.1 | AAP50497.1 | |||
| BC110894.1 | AAI10895.1 | |||
| BC139752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv550203 | Chr.10:21627539 - 21773800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map