Assay Details
Target Gene Details
Entrez Gene ID: | 55954 |
Gene Name: | zinc finger matrin-type 5 |
Gene Aliases: |
SNRNP20, U11/U12-20K, ZC3H19 |
Location: |
Chr.22:29730956-29767011 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMAT5 | NM_001003692.1 | NP_001003692.1 | ||
| NM_001318129.1 | NP_001305058.1 | |||
| NM_019103.2 | NP_061976.1 | |||
| AA312132.1 | ||||
| AK292671.1 | ||||
| AL160311.1 | CAB77284.1 | |||
| BC009717.1 | AAH09717.1 | |||
| BG285784.1 | ||||
| BQ931527.1 | ||||
| BU598307.1 | ||||
| CR456353.1 | CAG30239.1 | |||
| CR457393.1 | CAG33674.1 | |||
| DT217179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515720 | Chr.22:29685726 - 29767537 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map