Assay Details
Target Gene Details
Entrez Gene ID: | 3598 |
Gene Name: | interleukin 13 receptor subunit alpha 2 |
Gene Aliases: |
CD213A2, CT19, IL-13R, IL13BP |
Location: |
Chr.X:115003975-115017644 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL13RA2 | NM_000640.2 | 6 | 701 | NP_000631.1 |
| AK291957.1 | 6 | 670 | ||
| AK308523.1 | 6 | 632 | ||
| AK309444.1 | 6 | 711 | ||
| BC020739.1 | 5 | 602 | AAH20739.1 | |
| BC033705.2 | 5 | 610 | AAH33705.1 | |
| GQ494004.1 | 6 | 633 | ||
| U70981.1 | 6 | 669 | AAB17170.1 | |
| X95302.1 | 6 | 628 | CAA64617.1 | |
| Y08768.1 | 6 | 701 | CAA70021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516624 | Chr.X:114981600 - 115124568 on Build GRCh38 | Gain |
|
| esv3577049 | Chr.X:114971071 - 115158907 on Build GRCh38 | Gain |
|
| esv33994 | Chr.X:113685854 - 115738426 on Build GRCh38 | Gain+Loss |
|
| nsv513764 | Chr.X:114967659 - 115163287 on Build GRCh38 | Insertion |
|
| nsv511667 | Chr.X:114971055 - 115172427 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map