Assay Details
Target Gene Details
Entrez Gene ID: | 60496 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase |
Gene Aliases: |
AASD-PPT, ACPS, CGI-80, LYS2, LYS5 |
Location: |
Chr.11:106077565-106098692 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AASDHPPT | NM_015423.2 | NP_056238.2 | ||
| AF136978.1 | AAG49439.1 | |||
| AF151057.1 | AAF36143.1 | |||
| AF151838.1 | AAD34075.1 | |||
| AF201943.1 | AAF86879.1 | |||
| AF302110.1 | AAG30872.1 | |||
| AK293362.1 | ||||
| AK312529.1 | ||||
| AL050073.1 | CAB43257.1 | |||
| BC015470.1 | AAH15470.1 | |||
| BC016728.1 | AAH16728.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052674 | Chr.11:104111970 - 107043401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map