Assay Details
Target Gene Details
Entrez Gene ID: | 6324 |
Gene Name: | sodium voltage-gated channel beta subunit 1 |
Gene Aliases: |
ATFB13, BRGDA5, GEFSP1 |
Location: |
Chr.19:35030688-35040449 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN1B | NM_001037.4 | 6 | 1171 | NP_001028.1 |
| NM_001321605.1 | 6 | 1411 | NP_001308534.1 | |
| BC021266.2 | 5 | 942 | AAH21266.2 | |
| BC067122.1 | 6 | 1171 | AAH67122.1 | |
| L10338.1 | 6 | 1135 | AAA60391.1 | |
| L16242.1 | 6 | 1062 | AAA61277.1 |
More Information
Set Membership: |
|

Genomic Map