Assay Details
Target Gene Details
Entrez Gene ID: | 2244 |
Gene Name: | fibrinogen beta chain |
Gene Aliases: |
HEL-S-78p |
Location: |
Chr.4:154562980-154572763 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGB | NM_001184741.1 | 7 | 877 | NP_001171670.1 |
| NM_005141.4 | 7 | 1054 | NP_005132.2 | |
| AK303788.1 | 8 | 972 | ||
| AK309592.1 | 7 | 999 | ||
| AK312972.1 | 7 | 1054 | ||
| BC106760.2 | 7 | 1023 | AAI06761.1 | |
| BC107766.1 | 7 | 1023 | AAI07767.1 | |
| EU668333.1 | 7 | 1040 | ||
| J00129.1 | 7 | 991 | AAA52429.1 | |
| J00133.1 | 3 | 235 |
More Information
Set Membership: |
|

Genomic Map