Assay Details
Target Gene Details
Entrez Gene ID: | 131034 |
Gene Name: | copine 4 |
Gene Aliases: |
COPN4, CPN4 |
Location: |
Chr.3:131533560-132285702 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPNE4 | NM_001289112.1 | 16 | 2126 | NP_001276041.1 |
| NM_130808.2 | 16 | 2754 | NP_570720.1 | |
| NM_153429.1 | 17 | 2114 | NP_702907.1 | |
| XM_011512406.2 | 15 | 1901 | XP_011510708.1 | |
| XM_011512407.2 | 15 | 2158 | XP_011510709.1 | |
| XM_011512408.2 | 15 | 3286 | XP_011510710.1 | |
| XM_017005693.1 | 19 | 3507 | XP_016861182.1 | |
| XM_017005695.1 | 18 | 2377 | XP_016861184.1 | |
| XM_017005696.1 | 10 | 1271 | XP_016861185.1 | |
| AF465770.1 | AAL74190.1 | |||
| AF465771.1 | 17 | 2114 | AAL74191.1 | |
| AK056175.1 | 16 | 2226 | BAB71111.1 | |
| AK128117.1 | 20 | 4004 | ||
| AK295557.1 | ||||
| BC014396.1 | 16 | 2312 | AAH14396.1 | |
| BC028613.1 | 3 | 879 | AAH28613.1 | |
| BM930746.1 | 2 | 451 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006656 | Chr.3:131383932 - 132145572 on Build GRCh38 | Gain |
|
| nsv829724 | Chr.3:131366925 - 131557541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map