Assay Details
Target Gene Details
Entrez Gene ID: | 26061 |
Gene Name: | 2-hydroxyacyl-CoA lyase 1 |
Gene Aliases: |
2-HPCL, HPCL, HPCL2, PHYH2 |
Location: |
Chr.3:15560704-15601852 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACL1 | NM_001284413.1 | 16 | 2056 | NP_001271342.1 |
| NM_001284415.1 | 15 | 1957 | NP_001271344.1 | |
| NM_001284416.1 | 14 | 1891 | NP_001271345.1 | |
| NM_012260.3 | 17 | 2137 | NP_036392.2 | |
| NR_104315.1 | 15 | 1958 | ||
| AA643224.1 | 1 | 151 | ||
| AF161397.1 | 17 | 1757 | AAF28957.1 | |
| AJ131753.1 | 17 | 1847 | CAB60200.1 | |
| AK056977.1 | 9 | 1131 | ||
| AK301546.1 | 16 | 1773 | ||
| AK301990.1 | 14 | 1626 | ||
| AK302086.1 | 15 | 1674 | ||
| BC001627.1 | AAH01627.1 | |||
| BC007440.2 | 15 | 1663 | AAH07440.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834622 | Chr.3:15397807 - 15587083 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map