Assay Details
Target Gene Details
Entrez Gene ID: | 390892 |
Gene Name: | olfactory receptor family 7 subfamily A member 10 |
Gene Aliases: |
BC85395_3, OR19-18 |
Location: |
Chr.19:14840948-14841877 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7A10 | NM_001005190.1 | NP_001005190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061249 | Chr.19:14838727 - 14867472 on Build GRCh38 | Loss |
|
| esv2718220 | Chr.19:14840866 - 14852068 on Build GRCh38 | Deletion |
|
| nsv978778 | Chr.19:14840570 - 14846800 on Build GRCh38 | Duplication |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map