Assay Details
Target Gene Details
Entrez Gene ID: | 983 |
Gene Name: | cyclin dependent kinase 1 |
Gene Aliases: |
CDC2, CDC28A, P34CDC2 |
Location: |
Chr.10:60778331-60794852 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK1 | NM_001320918.1 | 5 | 537 | NP_001307847.1 |
| NM_001786.4 | 5 | 525 | NP_001777.1 | |
| NM_033379.4 | NP_203698.1 | |||
| XM_005270303.3 | 5 | 607 | XP_005270360.1 | |
| AK291939.1 | 5 | 462 | ||
| AW976143.1 | ||||
| BC014563.1 | 4 | 396 | AAH14563.1 | |
| BF685285.1 | 5 | 586 | ||
| BG723076.1 | 5 | 530 | ||
| BT007004.1 | 4 | 383 | AAP35650.1 | |
| CR933728.1 | 4 | 2756 | CAI46271.1 | |
| D88357.1 | BAA26001.1 | |||
| DA823877.1 | ||||
| X05360.1 | 5 | 509 | CAA28963.1 | |
| Y00272.1 | 5 | 509 | CAA68376.1 |
More Information
Set Membership: |
|

Genomic Map