Assay Details
Target Gene Details
Entrez Gene ID: | 116211 |
Gene Name: | transmembrane 4 L six family member 19 |
Gene Aliases: |
OCTM4 |
Location: |
Chr.3:196323546-196338420 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM4SF19 | NM_001204897.1 | 4 | 517 | NP_001191826.1 |
| NM_001204898.1 | 3 | 439 | NP_001191827.1 | |
| NM_138461.3 | 4 | 517 | NP_612470.2 | |
| AF192910.1 | 4 | 473 | AAQ13824.1 | |
| AK310209.1 | 4 | 487 | ||
| BC146985.1 | 4 | 418 | ||
| BC146993.1 | 4 | 418 | ||
| BC147021.1 | 4 | 418 | ||
| BC171870.1 | 3 | 340 | ||
| BP395549.1 | ||||
| BP396791.1 | ||||
| BQ678650.1 | 4 | 483 | ||
| CA430273.1 | 2 | 561 | ||
| CA489370.1 | 3 | 439 | ||
| DB015357.1 | 4 | 487 |
Target Gene Details
Entrez Gene ID: | 100874214 |
Gene Name: | TM4SF19 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:196318332-196325570 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM4SF19-AS1 | NR_046724.1 | 2 | 638 | |
| AK124973.1 | 2 | 620 |
Target Gene Details
Entrez Gene ID: | 100534611 |
Gene Name: | TM4SF19-TCTEX1D2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.3:196316085-196338420 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM4SF19-TCTEX1D2 | NR_037950.1 | 4 | 517 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997797 | Chr.3:195931489 - 196486087 on Build GRCh38 | Gain |
|
| nsv1009811 | Chr.3:196124925 - 196411200 on Build GRCh38 | Gain |
|
| nsv593071 | Chr.3:196132016 - 196345571 on Build GRCh38 | Gain |
|
| nsv4183 | Chr.3:196307196 - 196352238 on Build GRCh38 | Deletion |
|
| esv2759213 | Chr.3:195382058 - 196344722 on Build GRCh38 | Gain+Loss |
|

Genomic Map