Assay Details
Target Gene Details
Entrez Gene ID: | 282618 |
Gene Name: | interferon lambda 1 |
Gene Aliases: |
IL-29, IL29 |
Location: |
Chr.19:39296325-39298672 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNL1 | NM_172140.1 | 5 | 648 | NP_742152.1 |
| AY129150.1 | 5 | 648 | AAN28265.1 | |
| AY184372.1 | 5 | 551 | AAN86125.1 | |
| AY336716.1 | 5 | 554 | AAR24511.1 | |
| BC074985.2 | 5 | 571 | AAH74985.1 | |
| BC117482.1 | 5 | 596 | ||
| BC126183.1 | 5 | 596 | ||
| JF268782.1 | 5 | 551 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833830 | Chr.19:39218898 - 39347206 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map