Assay Details
Target Gene Details
Entrez Gene ID: | 26253 |
Gene Name: | C-type lectin domain family 4 member E |
Gene Aliases: |
CLECSF9, MINCLE |
Location: |
Chr.12:8533305-8541328 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC4E | NM_014358.3 | NP_055173.1 | ||
| XM_011520614.2 | XP_011518916.1 | |||
| XM_011520615.1 | XP_011518917.1 | |||
| AB024718.1 | BAA83755.1 | |||
| AK312671.1 | ||||
| AY358499.1 | AAQ88863.1 | |||
| BC000715.2 | AAH00715.1 | |||
| DA678013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv557342 | Chr.12:8446801 - 8621931 on Build GRCh38 | Loss |
|
| dgv1362n100 | Chr.12:8296504 - 8639973 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map