Assay Details
Target Gene Details
Entrez Gene ID: | 131450 |
Gene Name: | CD200 receptor 1 |
Gene Aliases: |
CD200R, HCRTR2, MOX2R, OX2R |
Location: |
Chr.3:112922685-112975090 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD200R1 | NM_138806.3 | NP_620161.1 | ||
| NM_170780.2 | NP_740750.1 | |||
| AF283760.1 | AAN61171.1 | |||
| AF495380.1 | AAM14622.1 | |||
| AF497548.1 | AAM16157.1 | |||
| AK126349.1 | ||||
| AK293071.1 | ||||
| AK304426.1 | ||||
| AY284975.1 | AAQ19772.1 | |||
| AY358910.1 | AAQ89269.1 | |||
| BC069661.1 | AAH69661.1 | |||
| BC069721.1 | AAH69721.1 | |||
| BC069743.1 | AAH69743.1 | |||
| BC093890.1 | AAH93890.1 | |||
| BC143393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597428 | Chr.3:112896541 - 112926195 on Build GRCh38 | Loss |
|
| nsv591293 | Chr.3:112266260 - 113293950 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113423961] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map