Assay Details
Target Gene Details
Entrez Gene ID: | 101927597 |
Gene Name: | NCK1 antisense RNA 1 (head to head) |
Gene Aliases: |
SLC35G2-AS1 |
Location: |
Chr.3:136841976-136862063 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCK1-AS1 | NR_110175.1 | |||
| NR_110176.1 | ||||
| NR_110177.1 | ||||
| CB047433.1 | ||||
| DA714434.1 | ||||
| HY007950.1 |
Target Gene Details
Entrez Gene ID: | 80723 |
Gene Name: | solute carrier family 35 member G2 |
Gene Aliases: |
TMEM22 |
Location: |
Chr.3:136819019-136855892 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35G2 | NM_001097599.1 | 2 | 1163 | NP_001091068.1 |
| NM_001097600.1 | 2 | 1014 | NP_001091069.1 | |
| NM_025246.2 | 2 | 1428 | NP_079522.2 | |
| XM_006713773.3 | 2 | 3580 | XP_006713836.1 | |
| XM_011513214.2 | 2 | 1767 | XP_011511516.1 | |
| XM_017007289.1 | 2 | 891 | XP_016862778.1 | |
| XM_017007290.1 | 2 | 1090 | XP_016862779.1 | |
| XM_017007291.1 | 2 | 896 | XP_016862780.1 | |
| AK314079.1 | 2 | 1323 | ||
| AL136661.1 | 2 | 1309 | CAB66596.1 | |
| BC000111.1 | 2 | 904 | AAH00111.1 | |
| BC022557.1 | 2 | 949 | AAH22557.1 |
More Information
Set Membership: |
|

Genomic Map