Assay Details
Target Gene Details
Entrez Gene ID: | 6524 |
Gene Name: | solute carrier family 5 member 2 |
Gene Aliases: |
SGLT2 |
Location: |
Chr.16:31482535-31490770 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A2 | NM_003041.3 | NP_003032.1 | ||
| NR_130783.1 | ||||
| XM_006721072.3 | XP_006721135.2 | |||
| AK054830.1 | 2 | 246 | ||
| BC131542.1 | ||||
| M95549.1 | AAA36608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060240 | Chr.16:31484552 - 31570129 on Build GRCh38 | Gain |
|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| nsv482193 | Chr.16:31483118 - 31490770 on Build GRCh38 | Gain |
|
| esv3570465 | Chr.16:31450067 - 31915765 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map