Assay Details
Target Gene Details
Entrez Gene ID: | 27199 |
Gene Name: | oxoglutarate receptor 1 |
Gene Aliases: |
GPR80, GPR99, P2RY15, P2Y15, aKGR |
Location: |
Chr.13:96985719-96995842 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXGR1 | NM_080818.3 | 4 | 2035 | NP_543008.3 |
| XM_005254043.3 | 4 | 2321 | XP_005254100.1 | |
| XM_005254044.3 | 4 | 3342 | XP_005254101.1 | |
| XM_005254045.2 | 4 | 2082 | XP_005254102.1 | |
| XM_005254046.2 | 4 | 2172 | XP_005254103.1 | |
| XM_011521077.2 | 4 | 2032 | XP_011519379.1 | |
| AK122639.1 | 4 | 2035 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832693 | Chr.13:96942591 - 97125227 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map