Assay Details
Target Gene Details
Entrez Gene ID: | 27339 |
Gene Name: | pre-mRNA processing factor 19 |
Gene Aliases: |
NMP200, PRP19, PSO4, SNEV, UBOX4, hPSO4 |
Location: |
Chr.11:60890547-60906589 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF19 | NM_014502.4 | 14 | 1399 | NP_055317.1 |
| AJ131186.1 | 14 | 1399 | CAB51857.1 | |
| BC008719.2 | 14 | 1394 | AAH08719.1 | |
| BC018665.2 | 14 | 1394 | AAH18665.1 | |
| BC018698.2 | 14 | 1394 | AAH18698.1 |
More Information
Set Membership: |
|

Genomic Map