Assay Details
Target Gene Details
Entrez Gene ID: | 5501 |
Gene Name: | protein phosphatase 1 catalytic subunit gamma |
Gene Aliases: |
PP-1G, PP1C, PPP1G |
Location: |
Chr.12:110708369-110742978 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1CC | NM_001244974.1 | NP_001231903.1 | ||
| NM_002710.3 | NP_002701.1 | |||
| XM_011538504.2 | XP_011536806.1 | |||
| XM_011538505.2 | XP_011536807.1 | |||
| AK297882.1 | ||||
| BC014073.2 | AAH14073.1 | |||
| DA472620.1 | ||||
| DA514523.1 | ||||
| L07395.1 | AAA19823.1 | |||
| X74008.1 | CAA52169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2675965 | Chr.12:110730751 - 110731820 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map