Assay Details
Target Gene Details
Entrez Gene ID: | 70 |
Gene Name: | actin, alpha, cardiac muscle 1 |
Gene Aliases: |
ACTC, ASD5, CMD1R, CMH11, LVNC4 |
Location: |
Chr.15:34788096-34795726 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTC1 | NM_005159.4 | 7 | 3177 | NP_005150.1 |
| AI222203.1 | ||||
| AK056592.1 | 7 | 2717 |
Target Gene Details
Entrez Gene ID: | 101928174 |
Gene Name: | uncharacterized LOC101928174 |
Gene Aliases: |
- |
Location: |
Chr.15:34755084-34812923 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928174 | NR_120329.1 | |||
| AK092087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569133 | Chr.15:34589103 - 34850365 on Build GRCh38 | Gain |
|
| esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain |
|
| nsv457106 | Chr.15:34757586 - 34935412 on Build GRCh38 | Gain |
|
| nsv522411 | Chr.15:34661785 - 35170780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map