Assay Details
Target Gene Details
Entrez Gene ID: | 55732 |
Gene Name: | chromosome 1 open reading frame 112 |
Gene Aliases: |
- |
Location: |
Chr.1:169794730-169854080 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf112 | NM_001320047.1 | NP_001306976.1 | ||
| NM_001320048.1 | NP_001306977.1 | |||
| NM_001320050.1 | NP_001306979.1 | |||
| NM_001320051.1 | NP_001306980.1 | |||
| NM_018186.3 | 1 | 185 | NP_060656.2 | |
| XM_005245317.4 | XP_005245374.1 | |||
| XM_017001722.1 | XP_016857211.1 | |||
| XM_017001723.1 | XP_016857212.1 | |||
| AK001568.1 | BAA91761.1 | |||
| AK096493.1 | ||||
| AK127098.1 | ||||
| AK299369.1 | ||||
| AK303296.1 | ||||
| AK310638.1 | ||||
| AL354614.1 | 1 | 185 | CAB89727.1 | |
| BC091516.1 | AAH91516.1 | |||
| DA770714.1 | ||||
| DC401980.1 |

Genomic Map